4-(Methylsulphonyl)benzaldehyde 95%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
424900025
ACRO424900025EA
104
EUR
ACRO424900025
4-(Methylsulphonyl)benzaldehyde 95%
4-(Methylsulphonyl)benzaldehyde
Appearance: Yellow to light beige to light brown Crystalline powder or crystals
Formel:
C₈H₈O₃S Schmelzpunkt: 155…161 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00216485 CAS-Nummer: 5398-77-6 |
Spezifikation Testergebnisse
Appearance | Yellow to light beige to light brown Crystalline powder or crystals |
Infrared spectrum | Conforms |
Melting point | 155 to 161 °C |
GC | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...