4-Methylsulphonylbenzylamine hydrochloride 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: (4-(Methylsulphonyl)phenyl)methanamine hydrochloride

424950025
ACRO424950025EA 106 EUR
ACRO424950025
4-Methylsulphonylbenzylamine hydrochloride 95%
4-Methylsulphonylbenzylamine hydrochloride

Appearance: White to beige to light yellow-pink Powder

Formel: C₈H₁₁NO₂S.HCl
Molecular Weight: 221,70 g/mol
Schmelzpunkt: 269…277 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00216491
CAS-Nummer: 98593-51-2

Bestellen

Spezifikation Testergebnisse

Appearance White to beige to light yellow-pink Powder
Infrared spectrum Conforms
Melting point 269 to 277 °C
Titration with NaOH ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR