1-Fluor-4-(methylsulfonyl)-2-nitrobenzol 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-Fluoro-5-methylsulphonylnitrobenzene, 1-Fluoro-4-(methylsulphonyl)-2-nitrobenzene

425000025
ACRO425000025EA 55.1 EUR
ACRO425000025
1-Fluor-4-(methylsulfonyl)-2-nitrobenzol 95%
1-Fluor-4-(methylsulfonyl)-2-nitrobenzol

Appearance: White to yellow-green Crystalline powder

Formel: C₇H₆FNO₄S
Molecular Weight: 219,19 g/mol
Schmelzpunkt: 152…158 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00025066
CAS-Nummer: 453-72-5

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow-green Crystalline powder
Infrared spectrum Conforms
Melting point 152 to 158 °C
HPLC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR