4'-Methoxy-2-chloracetophenon 97%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
2-Chloro-1-(4-methoxyphenyl)ethanone, 4-Methoxyphenacyl chloride
425260010
ACRO425260010EA
32.1
EUR
ACRO425260010
4'-Methoxy-2-chloracetophenon 97%
4'-Methoxy-2-chloracetophenon
Formel:
C₉H₉ClO₂ Molecular Weight: 184,6196 g/mol Schmelzpunkt: 98…100 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00216508 CAS-Nummer: 2196-99-8 UN: 1759 ADR: 8,II |
Spezifikation Testergebnisse
Appearance | Light yellow to beige Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 98 to 100 °C |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...