2-(4-Pyridyl)malondialdehyde 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-(Pyridin-4-yl)malonaldehyde

425340025
ACRO425340025EA 113 EUR
ACRO425340025
2-(4-Pyridyl)malondialdehyde 95%
2-(4-Pyridyl)malondialdehyde

Appearance: Beige to brown Powder

Formel: C₈H₇NO₂
Molecular Weight: 149,149 g/mol
Schmelzpunkt: >320 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00216535
CAS-Nummer: 51076-46-1

Bestellen

Spezifikation Testergebnisse

Appearance Beige to brown Powder
Infrared spectrum Conforms
Assay ≥94 %
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR