Methyl nicotinoylacetate 95%
Lieferant: Thermo Scientific
Synonyme:
Methyl 3-oxo-3-(pyridin-3-yl)propanoate
425490025
ACRO425490025EA
76.2
EUR
ACRO425490025
Methyl nicotinoylacetate 95%
Methyl nicotinoylacetate
Appearance: White to yellow to brown Crystalline powder or crystals and/or chunks
Formel:
C₉H₉NO₃ Molecular Weight: 179,175 g/mol Schmelzpunkt: 34…42 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00216524 CAS-Nummer: 54950-20-8 |
Spezifikation Testergebnisse
Appearance | White to yellow to brown Crystalline powder or crystals and/or chunks |
Infrared spectrum | Conforms |
Melting point | 34 to 42 °C |
HPLC | ≥94 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...