N-(4-Pentinyl)phthalimid 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-(Pent-4-yn-1-yl)isoindoline-1,3-dione
425730050
425730250
ACRO425730050EA
57.6
EUR
ACRO425730050
ACRO425730250
N-(4-Pentinyl)phthalimid 97%
N-(4-Pentinyl)phthalimid
Formel:
C₁₃H₁₁NO₂ Schmelzpunkt: 87…91 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06798110 CAS-Nummer: 6097-07-0 |
Spezifikation Testergebnisse
Appearance | White to brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 87 to 91 °C |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...