2-Fluor-m-anisaldehyd 96%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-Fluor-3-methoxybenzaldehyd, 2-Fluor-3-anisaldehyd
425830010
ACRO425830010EA
0
EUR
ACRO425830010
2-Fluor-m-anisaldehyd 96%
2-Fluor-m-anisaldehyd
Formel:
C₈H₇FO₂ Molecular Weight: 154,14 g/mol Schmelzpunkt: 47…51 °C Flammpunkt: >110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03411962 CAS-Nummer: 103438-88-6 |
Spezifikation Testergebnisse
Appearance | White to yellow Crystalline solid |
Infrared spectrum | Conforms |
Melting point | 44 to 48 °C |
GC | ≥95 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...