Benzyl-4-oxo-1-piperidinecarboxylate 97%

Lieferant: Thermo Fisher Scientific
Achtung

425960050 425960250
ACRO425960050EA 30.7 EUR
ACRO425960050 ACRO425960250
Benzyl-4-oxo-1-piperidinecarboxylate 97%
Benzyl-4-oxo-1-piperidinecarboxylate
Formel: C₁₃H₁₅NO₃
Siedepunkt: 114…140 °C (,3 mmHg)
Dichte: 1,185 g/cm³
Lagertemperatur: Raumtemperatur
MDL: MFCD00673144
CAS-Nummer: 19099-93-5

Bestellen

Spezifikation Testergebnisse

Appearance Colorless to white to light yellow to light orange Powder or solid or liquid
Infrared spectrum Conforms
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR