(2,3,6-Trifluorphenyl)essigsäure 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-(2,3,6-Trifluorophenyl)acetic acid

426030010
ACRO426030010EA 76 EUR
ACRO426030010
(2,3,6-Trifluorphenyl)essigsäure 98%
(2,3,6-Trifluorphenyl)essigsäure
Formel: C₆H₂F₃CH₂COOH
Schmelzpunkt: 111…114 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00061217
CAS-Nummer: 114152-23-7

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Crystalline powder or solid
Infrared spectrum Conforms
Melting point 111 to 115 °C
HPLC ≥97.5 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR