N-Boc-trans-4-(fmoc-amino)-L-proline 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
N-BOC-trans-4-N-FMOC-amino-L-proline, (2S,4R)-4-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-1-(tert-butoxycarbonyl)pyrrolidine-2-carboxylic acid
426040010
ACRO426040010EA
171
EUR
ACRO426040010
N-Boc-trans-4-(fmoc-amino)-L-proline 97%
N-Boc-trans-4-(fmoc-amino)-L-proline
Appearance: White Crystalline powder
Formel:
C₂₅H₂₈N₂O₆ Molecular Weight: 452,507 g/mol Schmelzpunkt: 199 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00673783 CAS-Nummer: 176486-63-8 |
Spezifikation Testergebnisse
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
NMR (1H-NMR) | consistent with structure |
Specific optical rotation | -33° to -35° (20°C, 589 nm) (c=1,chloroform) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...