4,7-Phenanthroline 98%

Lieferant: Thermo Fisher Scientific
Achtung

426100010
ACRO426100010EA 61.6 EUR
ACRO426100010
4,7-Phenanthroline 98%
4,7-Phenanthroline

Appearance: White to beige Crystalline powder

Formel: C₁₂H₈N₂
Molecular Weight: 180.21 g/mol
Schmelzpunkt: 172…174 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004987
CAS-Nummer: 230-07-9
UN: 2811
ADR: 6.1 III

Bestellen

Spezifikation Testergebnisse

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Melting point 170 to 174 °C
HPLC ≥97.5 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR