5-tert-Butyl-N-carbobenzoxy-L-glutamate 98+%

Lieferant: Thermo Scientific

Synonyme: Z-Glu(OtBu)-OH, N-Carbobenzoxy-L-glutamic Acid 5-tert-Butyl Ester, N-Cbz-L-glutamic Acid 5-tert-Butyl Ester, 5-tert-Butyl N-Cbz-L-glutamate

426150050
ACRO426150050EA 55.7 EUR
ACRO426150050
5-tert-Butyl-N-carbobenzoxy-L-glutamate 98+%
5-tert-Butyl-N-carbobenzoxy-L-glutamate

Appearance: White to off-white Powder

Formel: C₁₇H₂₃NO₆
Molecular Weight: 337,37 g/mol
Schmelzpunkt: 85 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00038274
CAS-Nummer: 3886-08-6

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 86 to 91 °C
HPLC ≥97.5 %
NMR (1H-NMR) Consistent with structure
Specific optical rotation -13° to -15° (20 °C, 589 nm) (c=2, CH3OH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR