4-(N-Boc-aminomethyl)aniline 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-(N-Boc-Aminomethyl)aniline

426200010
ACRO426200010EA 66.6 EUR
ACRO426200010
4-(N-Boc-aminomethyl)aniline 97%
4-(N-Boc-aminomethyl)aniline

Appearance: White to tan Crystalline powder

Formel: C₁₂H₁₈N₂O₂
Molecular Weight: 222,288 g/mol
Schmelzpunkt: 75…80 °C
Lagertemperatur: Kühlschrank
MDL: MFCD03001716
CAS-Nummer: 94838-55-8

Bestellen

Spezifikation Testergebnisse

Appearance White to tan Crystalline powder
Infrared spectrum Conforms
Melting point 73 to 81 °C
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR