2-(Diphenylphosphino)benzaldehyd 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: (2-Formylphenyl)diphenylphosphine

426400010
ACRO426400010EA 91 EUR
ACRO426400010
2-(Diphenylphosphino)benzaldehyd 97%
2-(Diphenylphosphino)benzaldehyde
Formel: C₁₉H₁₅OP
Molecular Weight: 290,30 g/mol
Schmelzpunkt: 112…115 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00013367
CAS-Nummer: 50777-76-9

Bestellen

Spezifikation Testergebnisse

Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Melting point 112 to 120 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR