L-Tyrosine disodium salt 98%

Lieferant: Thermo Fisher Scientific
Gefahr

426410250 426411000 426415000
ACRO426410250EA 59.1 EUR
ACRO426410250 ACRO426411000 ACRO426415000
L-Tyrosine disodium salt 98%
L-Tyrosine disodium salt

Appearance: White to tan Crystalline powder

Formel: C₁₈H₂₀N₂Na₂O₆
Schmelzpunkt: >300 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00037203
CAS-Nummer: 69847-45-6

Bestellen

Spezifikation Testergebnisse

Appearance White to tan Crystalline powder
Infrared spectrum Conforms
Water ≤19.3 %
HPLC ≥97.5 %
pH 10.5 to 11.5 (1 % in water)
Specific optical rotation -12.5° to -14.5° (20°C, 589 nm) (c=4 in H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR