(3S)-N,N-Dimethyl-3-pyrrolidinamin 97%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
(S)-3-(Dimethylamino)pyrrolidine, (S)-(-)-3-(Dimethylamino)pyrrolidine
426450010
ACRO426450010EA
143
EUR
ACRO426450010
(3S)-N,N-Dimethyl-3-pyrrolidinamin 97%
(3S)-(-)-3-(Dimethylamino)pyrrolidine
Formel:
C₆H₁₄N₂ Molecular Weight: 114,19 g/mol Siedepunkt: 164 °C (760 mmHg) Dichte: 0,899 g/cm³ Flammpunkt: 51 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00191348 CAS-Nummer: 132883-44-4 UN: 2733 ADR: 3,III |
Spezifikation Testergebnisse
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4640 to 1.4660 (20°C, 589 nm) |
GC | 96.0 % |
Specific optical rotation | -12° to -16° (20°C, 589 nm) (c=1% ethanol) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...