Bromoacetic anhydride 95%

Lieferant: Thermo Fisher Scientific
Gefahr

426700050 426700250
ACRO426700050EA 141 EUR
ACRO426700050 ACRO426700250
Bromoacetic anhydride 95%
Bromoacetic anhydride

Appearance: White to brown Solid

Formel: C₄H₄Br₂O₃
Lagertemperatur: Kühlschrank
MDL: MFCD00045041
CAS-Nummer: 13094-51-4
UN: 3265
ADR: 8,II

Bestellen

Spezifikation Testergebnisse

Appearance White to brown Solid
Infrared spectrum Conforms
Melting point 34 to 42 °C
Titration Argentometric ≥94 % (ex Br)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR