Ethyl aminohydroxyiminoacetate 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Ethyl 2-amino-2-(hydroxyimino)acetate, Ethyl 2-oximinooxamate
426880050
ACRO426880050EA
122
EUR
ACRO426880050
Ethyl aminohydroxyiminoacetate 98%
Ethyl aminohydroxyiminoacetate
Formel:
C₄H₈N₂O₃ Molecular Weight: 132,12 g/mol Schmelzpunkt: 99…103 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03013442 CAS-Nummer: 10489-74-4 |
Spezifikation Testergebnisse
Appearance | White to yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 96 to 103 °C |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...