2-Amino-4'-bromacetophenon Hydrochlorid 97%
Lieferant: Thermo Scientific
Synonyme:
4-Bromophenacylamine hydrochloride
427060050
ACRO427060050EA
85.6
EUR
ACRO427060050
2-Amino-4'-bromacetophenon Hydrochlorid 97%
2-Amino-4'-bromacetophenon Hydrochlorid
Formel:
C₈H₉BrClNO Molecular Weight: 250,53 g/mol Schmelzpunkt: 280…285 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00051998 CAS-Nummer: 5467-72-1 |
Spezifikation Testergebnisse
Appearance | Beige to pink-orange Powder |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
NMR (1H-NMR) | Consistent with structure |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...