N-Carbobenzoxy-1,3-diaminopropane hydrochloride 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: N-Benzyloxycarbonyl-1,3-propanediamine hydrochloride, N-Z-1,3-Propanediamine hydrochloride, N-Benzyloxycarbonyl-1,3-diaminopropane hydrochloride, N-Cbz-1,3-diaminopropane Hydrochloride, Benzyl N-(3-Aminopropyl)carbamate Hydrochloride, N-(3-Aminopropyl)carbamic Acid Benzyl Ester Hydrochloride, Benzyl (3-aminopropyl)carbamate hydrochloride

427070010 427070050
ACRO427070010EA 57.1 EUR
ACRO427070010 ACRO427070050
N-Carbobenzoxy-1,3-diaminopropane hydrochloride 98%
N-Carbobenzoxy-1,3-diaminopropane hydrochloride

Appearance: Yellow Crystalline powder

Formel: C₁₁H₁₆N₂O₂
Molecular Weight: 244.72 g/mol
Schmelzpunkt: 185…189 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00270153
CAS-Nummer: 17400-34-9

Bestellen

Spezifikation Testergebnisse

Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Melting point 185 to 189 °C
NMR consistent with structure
Titration with HClO4 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR