3,5-Dimethoxybenzylbromid 95%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
1-(Bromomethyl)-3,5-dimethoxybenzene
427340050
ACRO427340050EA
109
EUR
ACRO427340050
3,5-Dimethoxybenzylbromid 95%
3,5-Dimethoxybenzylbromid
Formel:
C₉H₁₁BrO₂ Molecular Weight: 231,0864 g/mol Schmelzpunkt: 69…70 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD01321368 CAS-Nummer: 877-88-3 UN: 3261 ADR: 8,II |
Spezifikation Testergebnisse
Appearance | White to brown Powder or crystals and/or chunks |
Infrared spectrum | Conforms |
Melting point | 68 to 72 °C |
HPLC | ≥94 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...