2-Indolcarbaldehyd 97%

Lieferant: Thermo Fisher Scientific
Achtung

427510010 427510050
ACRO427510010EA 109 EUR
ACRO427510010 ACRO427510050
2-Indolcarbaldehyd 97%
2-Indolcarbaldehyd
Formel: C₉H₇NO
Schmelzpunkt: 138…142 °C
Lagertemperatur: Kühlschrank
MDL: MFCD03001425
CAS-Nummer: 19005-93-7

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow to tan Crystalline powder
Infrared spectrum Conforms
Melting point 133 to 143 °C
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR