4-Hydroxy-3-methylbenzaldehyd 98%

Lieferant: Thermo Scientific
Achtung

427550010
ACRO427550010EA 93.3 EUR
ACRO427550010
4-Hydroxy-3-methylbenzaldehyd 98%
4-Hydroxy-3-methylbenzaldehyde
Formel: C₈H₈O₂
Molecular Weight: 136,1479 g/mol
Schmelzpunkt: 118…120 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012360
CAS-Nummer: 15174-69-3

Bestellen

Spezifikation Testergebnisse

Appearance Off-white to tan to brown Crystalline powder
Infrared spectrum Conforms
Melting point 118 to 120 °C
GC ≥97.5 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR