3-[(Benzyloxycarbonyl)amino]propionaldehyde 95%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
3-[(Benzyloxycarbonyl)amino]-1-propanal, Benzyl 3-oxopropylcarbamate, N-Phenylmethoxycarbonyl-3-aminopropanal
427720010
ACRO427720010EA
66.7
EUR
ACRO427720010
3-[(Benzyloxycarbonyl)amino]propionaldehyde 95%
3-[(Benzyloxycarbonyl)amino]propionaldehyde
Formel:
C₁₁H₁₃NO₃ Schmelzpunkt: 52…57 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD05664298 CAS-Nummer: 65564-05-8 |
Spezifikation Testergebnisse
Appearance | White to yellow Powder or solid |
Infrared spectrum | Conforms |
Melting point | 52 to 61 °C |
GC | ≥94 % |
NMR (1H-NMR) | Consistent with structure |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...