Bis(2-methyl-2-propanyl)-[(Z)-1H-pyrazol-1-ylmethylyliden]biscarbamat 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Bis-Boc-pyrazolocarboxamidine, 1-[N,N'-Bis(tert-butoxycarbonyl)carbamimidoyl]-1H-pyrazole, N,N'-Di-BOC-1H-pyrazole-1-carboxamidine, N,N'-Di-Boc-1H-pyrazole-1-carboximidamide, 1H-Pyrazole-1-carboxamidine, N,N'-Bis-BOC protected, N,N'-Bis(tert-butoxycarbonyl)-1H-pyrazole-1-carboximidamide

427970010 427970050 427970250
ACRO427970010EA 56.1 EUR
ACRO427970010 ACRO427970050 ACRO427970250
Bis(2-methyl-2-propanyl)-[(Z)-1H-pyrazol-1-ylmethylyliden]biscarbamat 98%
tert-Butyl (((tert-butoxycarbonyl)amino)(1H-pyrazol-1-yl)methylene)carbamate
Formel: C₁₄H₂₂N₄O₄
Schmelzpunkt: 86…90 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01075122
CAS-Nummer: 152120-54-2

Bestellen

Spezifikation Testergebnisse

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Melting point 86 to 90 °C
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR