L-(+)-Tartaric acid dipotassium salt 97%

Lieferant: Thermo Fisher Scientific

427991000 427995000
ACRO427991000EA 37 EUR
ACRO427991000 ACRO427995000
L-(+)-Tartaric acid dipotassium salt 97%
L-(+)-Tartaric acid dipotassium salt
Formel: C₄H₄K₂O₆
Lagertemperatur: Raumtemperatur
MDL: MFCD00013065
CAS-Nummer: 921-53-9

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤4 % (150°C, 4 hr)
Specific optical rotation +25° to +27° (20°C, 589 nm) (c=1,H2O)
Titration with HClO4 ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR