(R,R)-2,2'-Isopropylidenebis(4-phenyl-2-oxazoline)

Lieferant: Thermo Scientific
Achtung

Synonyme: (R,R)-2,2-Bis(4-phenyl-2-oxazolin-2-yl)propane, (R,R)-2,2-Isopropylidenebis(4-phenyl-2-oxazoline), (R,R)-2,2'-(Dimethylmethylene)bis(4-phenyl-2-oxazoline)

428050010
ACRO428050010EA 140 EUR
ACRO428050010
(R,R)-2,2'-Isopropylidenebis(4-phenyl-2-oxazoline)
(R,R)-2,2'-Isopropylidenebis(4-phenyl-2-oxazoline)

Appearance: Colorless to yellow viscous liquid or solid

Formel: C₂₁H₂₂N₂O₂
Molecular Weight: 334,42 g/mol
Schmelzpunkt: 56…58 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00192244
CAS-Nummer: 150529-93-4

Bestellen

Spezifikation Testergebnisse

Appearance Colorless to yellow viscous liquid or solid
Infrared spectrum Conforms
GC ≥96.0 %
Specific optical rotation +150° to +170° (20°C, 589 nm) (c=1%, ethanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR