2,2-Dimethyl-4-pentensäure 97%

Lieferant: Thermo Fisher Scientific
Gefahr

428090010
ACRO428090010EA 53 EUR
ACRO428090010
2,2-Dimethyl-4-pentensäure 97%
2,2-Dimethyl-4-pentenoic acid

Appearance: Clear colorless to yellow Liquid

Formel: C₇H₁₂O₂
Siedepunkt: 104…108 °C (20 mmHg)
Dichte: 0,933 g/cm³
Flammpunkt: 88 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012304
CAS-Nummer: 16386-93-9
UN: 3265
ADR: 8,II

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4310 to 1.4340 (20°C, 589 nm)
Titration with NaOH ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR