(S)-(+)-ɑ-(Trifluoromethyl)benzyl alcohol 99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
428180010
ACRO428180010EA
300
EUR
ACRO428180010
(S)-(+)-ɑ-(Trifluoromethyl)benzyl alcohol 99%
(S)-(+)-alpha-(Trifluoromethyl)benzyl alcohol
Appearance: Clear colorless to light yellow Liquid or low melting solid
Formel:
C₈H₇F₃O Siedepunkt: 127 °C (760 mmHg) Dichte: 1,3 g/cm³ Flammpunkt: 84 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00077845 CAS-Nummer: 340-06-7 UN: 2810 ADR: 6.1,III |
Spezifikation Testergebnisse
Appearance | Clear colorless to light yellow Liquid or low melting solid |
Infrared spectrum | Conforms |
Refractive index | 1.4610 to 1.4630 (20°C, 589 nm) |
GC | ≥98.5 % |
NMR | Consistent with structure |
Specific optical rotation | +30° to +33° (20°C, 589 nm) (C= neat) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...