1-Benzylindole-3-carboxylic acid 98%

Lieferant: Thermo Scientific

Synonyme: 1-Benzyl-1H-indole-3-carboxylic acid

428260050
ACRO428260050EA 210 EUR
ACRO428260050
1-Benzylindole-3-carboxylic acid 98%
1-Benzylindole-3-carboxylic acid
Formel: C₁₆H₁₃NO₂
Molecular Weight: 251,285 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00057094
CAS-Nummer: 27018-76-4

Bestellen

Spezifikation Testergebnisse

Appearance White to brown Crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR