EGTA-Na4 (Ethylenglycolbis(2-aminoethylether)-N,N,N',N'-tetraessigsäureTetranatriumsalz) 95%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
428570100
428570500
ACRO428570100EA
95
EUR
ACRO428570100
ACRO428570500
EGTA-Na4 (Ethylenglycolbis(2-aminoethylether)-N,N,N',N'-tetraessigsäureTetranatriumsalz) 95%
EGTA-Na4 (Ethylenglycolbis(2-aminoethylether)-N,N,N',N'-tetraessigsäureTetranatriumsalz)
Formel:
C₁₄H₂₀N₂Na₄O₁₀ Schmelzpunkt: 54…56 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00274420 CAS-Nummer: 13368-13-3 |
Spezifikation Testergebnisse
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Assay | 18.46 to 20.82 % (ICP on dried sample) |
Loss on drying | ≤20 % (110°C, 16 hrs) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...