N-α-Benzoyl-L-arginine 4-nitroanilide hydrochloride 98%

Lieferant: Thermo Scientific

Synonyme: Nα-Benzoyl-L-arginine 4-nitroanilide hydrochloride, Bz-Arg-pNA·HCl, Bz-Arg-PNA.HCl, N-α-Benzoyl-L-Arginine p-Nitroanilide Hydrochloride, (S)-N-(5-Guanidino-1-((4-nitrophenyl)amino)-1-oxopentan-2-yl)benzamide hydrochloride

428640010
ACRO428640010EA 581 EUR
ACRO428640010
N-α-Benzoyl-L-arginine 4-nitroanilide hydrochloride 98%
N-alpha-Benzoyl-L-arginine 4-nitroanilide hydrochloride
Formel: C₁₉H₂₃ClN₆O₄
Molecular Weight: 434,881 g/mol
Schmelzpunkt: 223 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00063682
CAS-Nummer: 21653-40-7

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow Powder
Infrared spectrum Conforms
HPLC ≥97.5 %
NMR (1H-NMR) Consistent with structure
Specific optical rotation +42° to +48° (20°C, 589 nm) (c=1 in DMF)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR