Isochinolin-8-amin 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Isoquinolin-8-amine, 8-Aminoisoquinoline

429400010 429400050
ACRO429400010EA 161 EUR
ACRO429400010 ACRO429400050
Isochinolin-8-amin 97%
Isochinolin-8-amin
Formel: C₉H₈N₂
Schmelzpunkt: 168…169 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00179553
CAS-Nummer: 23687-27-6
UN: 2811
ADR: 6.1,III

Bestellen

Spezifikation Testergebnisse

Appearance Brown Crystalline powder or solid
Infrared spectrum Conforms
Melting point 167 to 170 °C
GC ≥96.0 %
Residue after evaporation ≤0.0003 %
Conductivity ≤1 µS/cm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR