5-Chinoxalincarbonsäure 95%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
429580010
ACRO429580010EA
184
EUR
ACRO429580010
5-Chinoxalincarbonsäure 95%
5-Quinoxalinecarboxylic acid
Formel:
C₉H₆N₂O₂ Schmelzpunkt: 167,6…171.7 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD02854494 CAS-Nummer: 6924-66-9 |
Spezifikation Testergebnisse
Appearance | Brown Powder |
Infrared spectrum | Conforms |
Melting point | 166 to 173 °C |
HPLC | ≥94.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...