1H-Indazol-7-carbonsäure 97%

Lieferant: Thermo Scientific
Achtung

429600010
ACRO429600010EA 184 EUR
ACRO429600010
1H-Indazol-7-carbonsäure 97%
1H-Indazole-7-carboxylic acid
Formel: C₈H₆N₂O₂
Molecular Weight: 162,15 g/mol
Schmelzpunkt: 229…231 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06804572
CAS-Nummer: 677304-69-7

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow Crystalline powder
Infrared spectrum Conforms
Melting point 228 to 233 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR