1,1-Dimethylbiguanid Hydrochlorid 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
429720050
ACRO429720050EA
78.5
EUR
ACRO429720050
1,1-Dimethylbiguanid Hydrochlorid 97%
1,1-Dimethylbiguanid Hydrochlorid
Formel:
C₄H₁₂ClN₅ Schmelzpunkt: 218…223 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012582 CAS-Nummer: 1115-70-4 |
Spezifikation Testergebnisse
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 216 to 224 °C |
HPLC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...