2-Brom-5-thiazolcarbonsäure 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-Bromothiazole-5-carboxylic acid
429760010
429760050
ACRO429760010EA
65.6
EUR
ACRO429760010
ACRO429760050
2-Brom-5-thiazolcarbonsäure 97%
2-Bromo-5-thiazolecarboxylic acid
Formel:
C₄H₂BrNO₂S Schmelzpunkt: 174,5…175.5 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04115730 CAS-Nummer: 54045-76-0 |
Spezifikation Testergebnisse
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 173 to 177 °C |
HPLC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...