2-Brom-5-thiazolcarbonsäure 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-Bromothiazole-5-carboxylic acid

429760010 429760050
ACRO429760010EA 65.6 EUR
ACRO429760010 ACRO429760050
2-Brom-5-thiazolcarbonsäure 97%
2-Bromo-5-thiazolecarboxylic acid
Formel: C₄H₂BrNO₂S
Schmelzpunkt: 174,5…175.5 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04115730
CAS-Nummer: 54045-76-0

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 173 to 177 °C
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR