2-Pyrimidincarbonitril 98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 2-Cyanopyrimidine

429820050 429820250
ACRO429820050EA 35.1 EUR
ACRO429820050 ACRO429820250
2-Pyrimidincarbonitril 98%
2-Pyrimidincarbonitril
Formel: C₅H₃N₃
Molecular Weight: 105.1 g/mol
Schmelzpunkt: 37…41 °C
Flammpunkt: 110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00160513
CAS-Nummer: 14080-23-0
UN: 3439
ADR: 6.1 III

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder or low melting solid
Infrared spectrum Conforms
Melting point 35 to 42 °C
GC ≥97.5 %
Specific optical rotation +96° to +107° (20 °C, 589 nm) (C=1, EtOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR