3-Brom-4-chlorbenzoesäure 97%

Lieferant: Thermo Scientific
Achtung

429870050
ACRO429870050EA 186 EUR
ACRO429870050
3-Brom-4-chlorbenzoesäure 97%
3-Bromo-4-chlorobenzoic acid
Formel: C₇H₄BrClO₂
Molecular Weight: 235,46 g/mol
Schmelzpunkt: 220…222 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00079706
CAS-Nummer: 42860-10-6

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 219 to 223 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR