3-Brom-5-hydroxypyridin 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 3-Brom-5-pyridinol

429900010
ACRO429900010EA 41.3 EUR
ACRO429900010
3-Brom-5-hydroxypyridin 97%
3-Brom-5-hydroxypyridin
Formel: C₅H₄BrNO
Molecular Weight: 174 g/mol
Schmelzpunkt: 168…169,5 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00661305
CAS-Nummer: 74115-13-2
ADR: 6.1,III

Bestellen

Spezifikation Testergebnisse

Appearance White to tan Crystalline powder
Infrared spectrum Conforms
Melting point 167 to 171 °C
HPLC ≥96.0 %
Sulfide (S) ≤0.0002 %
Carbonate (CO3) ≤0.001 %
Phosphate (PO4) ≤0.00005 %
Sulfate (SO4) ≤0.0002 %
Barium (Ba) ≤0.00001 %
Bismuth (Bi) ≤0.00001 %
Cadmium (Cd) ≤0.000005 %
Calcium (Ca) ≤0.0001 %
Chromium (Cr) ≤0.000005 %
Cobalt (Co) ≤0.00001 %
Copper (Cu) ≤0.00001 %
Gallium (Ga) ≤0.000002 %
Indium (In) ≤0.000002 %
Iron (Fe) ≤0.00001 %
Lead (Pb) ≤0.000005 %
Lithium (Li) ≤0.000002 %
Magnesium (Mg) ≤0.00005 %
Manganese (Mn) ≤0.00001 %
Molybdenum (Mo) ≤0.00001 %
Nickel (Ni) ≤0.000005 %
Platinum (Pt) ≤0.00001 %
Potassium (K) ≤0.0001 %
Sodium (Na) ≤0.0001 %
Strontium (Sr) ≤0.0001 %
Thallium (Tl) ≤0.000005 %
Tin (Sn) ≤0.00001 %
Titanium (Ti) ≤0.00001 %
Zinc (Zn) ≤0.00001 %
Reducing substances ≤0.0005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR