3-Chlorpicolinsäure 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3-Chloropyridine-2-carboxylic acid
429912500
ACRO429912500EA
77.8
EUR
ACRO429912500
3-Chlorpicolinsäure 97%
3-Chlorpicolinsäure
Formel:
C₆H₄ClNO₂ Molecular Weight: 157,56 g/mol Schmelzpunkt: 124…126 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01646325 CAS-Nummer: 57266-69-0 |
Spezifikation Testergebnisse
Appearance | White to brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 124 to 127 °C |
HPLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...