Resveratrol 99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
trans-3,4,5-Trihydroxystilbene, (E)-5-(4-Hydroxystyryl)benzene-1,3-diol
430075000
ACRO430075000EA
471
EUR
ACRO430075000
Resveratrol 99%
Resveratrol
Formel:
C₁₄H₁₂O₃ Molecular Weight: 228.25 g/mol Schmelzpunkt: 261…263 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00133799 CAS-Nummer: 501-36-0 Merck Index: 12,08326 |
Spezifikation Testergebnisse
Appearance | Off-white to beige Powder |
Infrared spectrum | Conforms |
Melting point | 257 to 264 °C |
Loss on drying | ≤1 % |
HPLC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...