1H-Indazol-4-carbonsäure 97%

Lieferant: Thermo Fisher Scientific
Achtung

430090010
ACRO430090010EA 139 EUR
ACRO430090010
1H-Indazol-4-carbonsäure 97%
1H-Indazole-4-carboxylic acid
Formel: C₈H₆N₂O₂
Molecular Weight: 162.15 g/mol
Schmelzpunkt: 209.5…211 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03840644
CAS-Nummer: 677306-38-6

Bestellen

Spezifikation Testergebnisse

Appearance White to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 207 to 212 °C
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR