4-Nitro-1H-pyrazol 97%

Lieferant: Thermo Scientific
Achtung

430160050
ACRO430160050EA 49.8 EUR
ACRO430160050
4-Nitro-1H-pyrazol 97%
4-Nitro-1H-pyrazole
Formel: C₃H₃N₃O₂
Molecular Weight: 113,0800 g/mol
Schmelzpunkt: 161…162 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00159626
CAS-Nummer: 2075-46-9

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 160 to 165 °C
HPLC ≥96.0 %
UV transmittance ≥99 % (c=0.01M water, 254 nm)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR