4-Methyl-3-pyridinboronsäure 90%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: (4-Methylpyridin-3-yl)boronic acid, 4-Methylpyridine-3-boronic acid

430270010
ACRO430270010EA 122 EUR
ACRO430270010
4-Methyl-3-pyridinboronsäure 90%
4-Methyl-3-pyridineboronic acid
Formel: C₆H₈BNO₂
Molecular Weight: 136.94 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03788244
CAS-Nummer: 148546-82-1

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
HPLC ≥88.0 %
Residue after evaporation ≤0.0005 %
Acidity ≤0.0002 meq/g
Substance darkened by H2SO4 Passes test
Aluminium (Al) ≤0.5 ppm
Barium (Ba) ≤0.1 ppm
Cadmium (Cd) ≤0.05 ppm
Calcium (Ca) ≤0.5 ppm
Chromium (Cr) ≤0.02 ppm
Copper (Cu) ≤0.02 ppm
Iron (Fe) ≤0.1 ppm
Lead (Pb) ≤0.1 ppm
Magnesium (Mg) ≤0.1 ppm
Manganese (Mn) ≤0.02 ppm
Nickel (Ni) ≤0.02 ppm
Zinc (Zn) ≤0.1 ppm
Sulfur compounds (as S) ≤0.005 %
Aromatics ≤0.01 % Benzene

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR