6-Benzyloxy-1-boc-indole-2-boronic acid 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
1H-Indole-1-carboxylicacid, 2-borono-6-(phenylmethoxy)-, 1,1-dimethylethyl ester, 6-Benzyloxy-1H-indole-2-boronic acid, N-BOC protected, 1-(TERT-BUTOXYCARBONYL)-6-(BENZYLOXY)-1H-INDOL-2-YL-2-BORONIC ACID
430330010
ACRO430330010EA
158
EUR
ACRO430330010
6-Benzyloxy-1-boc-indole-2-boronic acid 97%
6-Benzyloxy-1-boc-indole-2-boronic acid
Formel:
C₂₀H₂₂BNO₅ Schmelzpunkt: 102…106 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD06659831 CAS-Nummer: 850568-66-0 |
Spezifikation Testergebnisse
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 101 to 106 °C |
HPLC | ≥96.0 % |
Acidity | ≤0.0005 meq/g |
Chloride (Cl) | ≤0.001 % |
Sulfate (SO4) | ≤0.001 % |
Cadmium (Cd) | ≤0.05 ppm |
Calcium (Ca) | ≤0.5 ppm |
Chromium (Cr) | ≤0.02 ppm |
Copper (Cu) | ≤0.02 ppm |
Iron (Fe) | ≤0.1 ppm |
Lead (Pb) | ≤0.1 ppm |
Magnesium (Mg) | ≤0.1 ppm |
Nickel (Ni) | ≤0.02 ppm |
Zinc (Zn) | ≤0.1 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...