2-Chlor-3-fluorphenylboronsäure 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-Chloro-3-fluorobenzeneboronic acid, (2-Chloro-3-fluorophenyl)boronic acid
430370010
ACRO430370010EA
169
EUR
ACRO430370010
2-Chlor-3-fluorphenylboronsäure 97%
2-Chlor-3-fluorphenylboronsäure
Formel:
C₆H₅BClFO₂ Schmelzpunkt: 115,8…115.9 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD07363741 CAS-Nummer: 871329-52-1 |
Spezifikation Testergebnisse
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 113 to 117 °C |
HPLC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...