2-Methyl-3-pyridinboronsäure Hydrochlorid 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-Methylpyridine-3-boronic acid hydrochloride, (2-Methylpyridin-3-yl)boronic acid hydrochloride
430390010
ACRO430390010EA
200
EUR
ACRO430390010
2-Methyl-3-pyridinboronsäure Hydrochlorid 97%
2-Methyl-3-pyridinboronsäure Hydrochlorid
Formel:
C₆H₉BClNO₂ Schmelzpunkt: 181…186 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06659489 CAS-Nummer: 1072952-34-1 |
Spezifikation Testergebnisse
Appearance | Off-white Crystalline solid |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Sulfate (SO4) | ≤0.1 % |
Iron (Fe) | ≤0.01 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...